ENA497681910 Enamine

3-(2,6-difluorophenyl)propan-1-ol

  • MDL number MFCD09028680
  • Cas Number : 401939-88-6
  • Molecular Weight : 172.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266308
Availability details for
Quantity
  • Availability
  • Add to Cart