ENA497681933 Enamine

ethyl 5-iodo-1,3-thiazole-2-carboxylate

  • MDL number MFCD21728858
  • Cas Number : 1500358-48-4
  • Molecular Weight : 283.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266371
Availability details for
Quantity
  • Availability
  • Add to Cart