ENA497681938 Enamine

2-chloro-5-(propan-2-yl)-1,3-benzoxazole

  • MDL number MFCD11890744
  • Cas Number : 155392-52-2
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266379
Availability details for
Quantity
  • Availability
  • Add to Cart