ENA497681942 Enamine

1-(4-fluorophenyl)-3-methylbut-2-en-1-one

  • MDL number MFCD21818220
  • Cas Number : 92236-75-4
  • Molecular Weight : 178.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266386
Availability details for
Quantity
  • Availability
  • Add to Cart