ENA497681947 Enamine

(4-fluorocyclohexyl)methanesulfonyl chloride

  • MDL number MFCD28547239
  • Cas Number : 1783346-01-9
  • Molecular Weight : 214.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266398
Availability details for
Quantity
  • Availability
  • Add to Cart