ENA497681950 Enamine

4,5-dimethyloxolan-3-one

  • MDL number MFCD29907173
  • Cas Number : 90238-77-0
  • Molecular Weight : 114.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266405
Availability details for
Quantity
  • Availability
  • Add to Cart