ENA497681965 Enamine

[(4-{[(2R,6S)-2,6-dimethylmorpholin-4-yl]methyl}phenyl)methyl](methyl)amine

  • MDL number MFCD29907181
  • Cas Number : 1909287-67-7
  • Molecular Weight : 248.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266465
Availability details for
Quantity
  • Availability
  • Add to Cart