ENA497681986 Enamine

N-(prop-2-en-1-yl)-1,2,3,4-thiatriazol-5-amine

  • MDL number MFCD11850874
  • Cas Number : 77695-07-9
  • Molecular Weight : 142.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266542
Availability details for
Quantity
  • Availability
  • Add to Cart