ENA497682018 Enamine

2-phenyl-2-(phenylamino)ethan-1-ol

  • MDL number MFCD12826217
  • Cas Number : 13891-02-6
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266581
Availability details for
Quantity
  • Availability
  • Add to Cart