ENA497682055 Enamine

5-(iodomethyl)-2-phenyl-4,5-dihydro-1,3-oxazole

  • MDL number MFCD29907197
  • Cas Number : 200573-05-3
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266643
Availability details for
Quantity
  • Availability
  • Add to Cart