ENA497682059 Enamine

1-(pyridin-3-yl)-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD07381325
  • Cas Number : 342601-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266665
Availability details for
Quantity
  • Availability
  • Add to Cart