ENA497682060 Enamine

2-chloro-4-(3,3-dimethylpiperidin-1-yl)pyrimidine

  • MDL number MFCD09750071
  • Cas Number : 954227-28-2
  • Molecular Weight : 225.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266685
Availability details for
Quantity
  • Availability
  • Add to Cart