ENA497682067 Enamine

bis[(propan-2-yl)amino]phosphinoyl fluoride

  • MDL number MFCD00801119
  • Cas Number : 371-86-8
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-62
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266699

Safety Information

signalword Danger
pcodes P260,P264,P270,P301+P310,P307+P311,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-266699
pictograms GHS06,GHS08
hstatements H301,H370
Availability details for
Quantity
  • Availability
  • Add to Cart