ENA497682078 Enamine

benzyl (2S,3aR,7aS)-octahydro-1H-indole-2-carboxylate hydrochloride

  • MDL number MFCD09750985
  • Cas Number : 145641-35-6
  • Molecular Weight : 295.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266719
Availability details for
Quantity
  • Availability
  • Add to Cart