ENA497682082 Enamine

4-(1,3-dioxolan-2-yl)benzaldehyde

  • MDL number MFCD03180272
  • Cas Number : 40681-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-266737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266737
Availability details for
Quantity
  • Availability
  • Add to Cart