ENA497682102 Enamine

2-[(2-bromophenyl)amino]ethan-1-ol

  • MDL number MFCD11153106
  • Cas Number : 87762-20-7
  • Molecular Weight : 216.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-266858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-266858
Availability details for
Quantity
  • Availability
  • Add to Cart