ENA497682143 Enamine

5-[(benzenesulfonyl)methyl]-2-chloro-1,3-thiazole

  • MDL number MFCD01568640
  • Cas Number : 320423-60-7
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267009
Availability details for
Quantity
  • Availability
  • Add to Cart