ENA497682149 Enamine

methyl 5-[2-(trimethylsilyl)ethynyl]pyridine-2-carboxylate

  • MDL number MFCD16036471
  • Cas Number : 216444-02-9
  • Molecular Weight : 233.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267025
Availability details for
Quantity
  • Availability
  • Add to Cart