ENA497682183 Enamine

3-isothiocyanato-N,N-dimethylpropanamide

  • MDL number MFCD16125995
  • Cas Number : 1248198-30-2
  • Molecular Weight : 158.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267101
Availability details for
Quantity
  • Availability
  • Add to Cart