ENA497682204 Enamine

1-(pyrrolidin-1-yl)propan-2-ol

  • MDL number MFCD00020854
  • Cas Number : 42122-41-8
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267163
Availability details for
Quantity
  • Availability
  • Add to Cart