ENA497682207 Enamine

2-(2,2-difluoro-1,3-dioxaindan-5-yl)ethan-1-ol

  • MDL number MFCD09926421
  • Cas Number : 641631-96-1
  • Molecular Weight : 202.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267176
Availability details for
Quantity
  • Availability
  • Add to Cart