ENA497682208 Enamine

4-(3,4-dimethylphenyl)oxane-4-carboxylic acid

  • MDL number MFCD17141003
  • Cas Number : 1489920-11-7
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267177
Availability details for
Quantity
  • Availability
  • Add to Cart