ENA497682227 Enamine

butyl 2-amino-2-methylpropanoate hydrochloride

  • MDL number MFCD29907258
  • Cas Number : 1909317-21-0
  • Molecular Weight : 195.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267232
Availability details for
Quantity
  • Availability
  • Add to Cart