ENA497682249 Enamine

3-(naphthalen-1-yl)azetidin-3-ol hydrochloride

  • MDL number MFCD29907268
  • Cas Number : 1909313-76-3
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267297
Availability details for
Quantity
  • Availability
  • Add to Cart