ENA497682254 Enamine

dimethyl (cyanomethyl)phosphonate

  • MDL number MFCD00039486
  • Cas Number : 21658-92-4
  • Molecular Weight : 149.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267326

Safety Information

signalword Danger
pcodes P201,P202,P261,P264,P271,P280,P281,P304+P340,P305+P351+P338,P308+P313,P312,P337+P313,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-267326
pictograms GHS07,GHS08
hstatements H319,H332,H340
Availability details for
Quantity
  • Availability
  • Add to Cart