ENA497682259 Enamine

6-amino-5-[2-(hydroxymethyl)morpholin-4-yl]-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD23351139
  • Cas Number : 1511744-91-4
  • Molecular Weight : 256.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267332
Availability details for
Quantity
  • Availability
  • Add to Cart