ENA497682272 Enamine

3-isothiocyanato-N-methylpropanamide

  • MDL number MFCD16125998
  • Cas Number : 63767-54-4
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267352
Availability details for
Quantity
  • Availability
  • Add to Cart