ENA497682287 Enamine

(1R)-1-(2-fluorophenyl)propan-1-amine hydrochloride

  • MDL number MFCD12757043
  • Cas Number : 1168139-44-3
  • Molecular Weight : 189.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267374
Availability details for
Quantity
  • Availability
  • Add to Cart