ENA497682305 Enamine

2-chloro-N-[dimethyl(oxo)-lambda6-sulfanylidene]acetamide

  • MDL number MFCD29907284
  • Cas Number : 1855705-79-1
  • Molecular Weight : 169.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267469
Availability details for
Quantity
  • Availability
  • Add to Cart