ENA497682315 Enamine

5-chloro-6-[(3-chlorophenyl)methoxy]pyridine-3-carboxylic acid

  • MDL number MFCD21480813
  • Cas Number : 1489854-56-9
  • Molecular Weight : 298.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267495
Availability details for
Quantity
  • Availability
  • Add to Cart