ENA497682316 Enamine

2-[(2R,6S)-2,6-dimethylmorpholin-4-yl]propan-1-amine

  • MDL number MFCD28149646
  • Cas Number : 1592018-91-1
  • Molecular Weight : 172.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267507
Availability details for
Quantity
  • Availability
  • Add to Cart