ENA497682320 Enamine

N-ethyl-1-methyl-1H-pyrazol-4-amine

  • MDL number MFCD12195615
  • Cas Number : 1084976-66-8
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267518

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-267518
pictograms GHS07
hstatements H302,H312
Availability details for
Quantity
  • Availability
  • Add to Cart