ENA497682329 Enamine

1-(4-aminopiperidin-1-yl)-2,2-dimethylpropan-1-one hydrochloride

  • MDL number MFCD17014374
  • Cas Number : 1286264-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-267530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267530
Availability details for
Quantity
  • Availability
  • Add to Cart