ENA497682338 Enamine

ethyl 5-cyanopentanoate

  • MDL number MFCD29907296
  • Cas Number : 4450-39-9
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267557
Availability details for
Quantity
  • Availability
  • Add to Cart