ENA497682354 Enamine

2-nitro-5-(trifluoromethyl)furan

  • MDL number MFCD03002311
  • Cas Number : 1524-31-8
  • Molecular Weight : 181.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267639
Availability details for
Quantity
  • Availability
  • Add to Cart