ENA497682377 Enamine

2,4-difluoro-6-methylpyrimidine

  • MDL number MFCD18377655
  • Cas Number : 696-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-267689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267689
Availability details for
Quantity
  • Availability
  • Add to Cart