ENA497682542 Enamine

(3R)-2-methylpentan-3-amine hydrochloride

  • MDL number MFCD29907312
  • Cas Number : 196929-99-4
  • Molecular Weight : 137.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267753
Availability details for
Quantity
  • Availability
  • Add to Cart