ENA497682568 Enamine

2-tert-butyl-1-methylpiperazine dihydrochloride

  • MDL number MFCD29907346
  • Cas Number : 1909347-65-4
  • Molecular Weight : 229.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268142
Availability details for
Quantity
  • Availability
  • Add to Cart