ENA497682603 Enamine

3-(1-methylpyrrolidin-2-yl)-1H-pyrazole-5-carboxylic acid hydrochloride

  • MDL number MFCD29907382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-268506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268506
Availability details for
Quantity
  • Availability
  • Add to Cart