ENA497682604 Enamine

methyl 5-amino-1,3-dimethyl-2,4-dioxo-1H,2H,3H,4H-thieno[2,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD29907383
  • Cas Number : 1909316-25-1
  • Molecular Weight : 269.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268510
Availability details for
Quantity
  • Availability
  • Add to Cart