ENA497682637 Enamine

3-(5-chloro-1,2,3-thiadiazol-4-yl)aniline hydrochloride

  • MDL number MFCD29907419
  • Cas Number : 1909316-42-2
  • Molecular Weight : 248.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269407
Availability details for
Quantity
  • Availability
  • Add to Cart