ENA497682682 Enamine

1-[2-(tert-butoxy)-2-oxoethyl]-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD29907470
  • Cas Number : 1909336-14-6
  • Molecular Weight : 225.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-98139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98139
Availability details for
Quantity
  • Availability
  • Add to Cart