ENA497682706 Enamine

6-amino-N,N-dimethylpyridazine-3-carboxamide

  • MDL number MFCD16688695
  • Cas Number : 1250216-83-1
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268364
Availability details for
Quantity
  • Availability
  • Add to Cart