ENA497682708 Enamine

{4H,6H,7H-pyrazolo[3,2-c][1,4]oxazin-2-yl}methanol

  • MDL number MFCD16620641
  • Cas Number : 623565-58-2
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6,7-dihydro-4H-pyrazolo[5,1-c][1,4]oxazin-2-ylmethanol
Inchi Code 1S/C7H10N2O2/c10-4-6-3-7-5-11-2-1-9(7)8-6/h3,10H,1-2,4-5H2
Inchi Key BHXIVJNUCBWSFO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267889

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-267889
pictograms GHS07
hstatements H303,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart