ENA497682710 Enamine

4-chloro-5-methoxy-6-methylpyrimidine

  • MDL number MFCD24154252
  • Cas Number : 1739-60-2
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267890
Availability details for
Quantity
  • Availability
  • Add to Cart