ENA497682712 Enamine

6-(chloromethyl)-2-methyl-1,3-benzothiazole

  • MDL number MFCD18826618
  • Cas Number : 99846-82-9
  • Molecular Weight : 197.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267891
Availability details for
Quantity
  • Availability
  • Add to Cart