ENA497682728 Enamine

(3-bromo-2-methylphenyl)methanesulfonamide

  • MDL number MFCD26064757
  • Cas Number : 1567106-72-2
  • Molecular Weight : 264.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268480
Availability details for
Quantity
  • Availability
  • Add to Cart