ENA497682746 Enamine

(1S)-1-(1-methyl-1H-1,2,3-triazol-5-yl)ethan-1-ol

  • MDL number MFCD29907318
  • Cas Number : 1846687-41-9
  • Molecular Weight : 127.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267812
Availability details for
Quantity
  • Availability
  • Add to Cart