ENA497682751 Enamine

2,7-dioxaspiro[4.4]nonane-1,3-dione

  • MDL number MFCD18332666
  • Cas Number : 1343062-15-6
  • Molecular Weight : 156.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-71
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267979
Availability details for
Quantity
  • Availability
  • Add to Cart