ENA497682760 Enamine

2-chloro-1-(1-methyl-1H-pyrazol-5-yl)ethan-1-one

  • MDL number MFCD11847299
  • Cas Number : 1260878-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-268537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268537
Availability details for
Quantity
  • Availability
  • Add to Cart